Learn More
Medchemexpress LLC HY-100612 5mg Medchemexpress, T16Ainh-A01 CAS:552309-42-9 Purity:>98%
SDP
Catalog No. 501967758
Encompass_Preferred
Medchemexpress, HY-100612 5mg T16Ainh-A01 CAS:552309-42-9 T16Ainh-A01, an aminophenylthiazole, is a potent transmembrane protein 16A (TMEM16A) inhibitor, inhibiting TMEM16A-mediated chloride currents with an IC50 value of ~1 µM. TMEM16A (ANO1) functions as a calcium-activated chloride channel (CaCC). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7758 Supplier Medchemexpress LLC Supplier No. HY1006125MG
Medchemexpress, HY-100612 5mg T16Ainh-A01 CAS:552309-42-9 T16Ainh-A01, an aminophenylthiazole, is a potent transmembrane protein 16A (TMEM16A) inhibitor, inhibiting TMEM16A-mediated chloride currents with an IC50 value of ~1 µM. TMEM16A (ANO1) functions as a calcium-activated chloride channel (CaCC). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.