Learn More
Sarstedt Inc T175 FLASK CELL+ CN VENT/CS40
Supplier:  Sarstedt Inc 83.3912.302
Encompass_Preferred
T175 Tissue Culture Flask Cell+ Canted Neck Yellow Ventilated Cap for Primary and Adherent Cells
Catalog No. NC9119710