Learn More
ATCC T2; Lymphoblast; Human (Homo sapiens)
Catalog No. 502382070
Encompass
T2 (174 x CEM.T2) cellls were derived from a variant of the T1 cell line (ATCC CRL-1991) produced by selection the SFR1-MI.3 monoclonal antibody (against a monomorphic determinant on HLA DR). The cell line can be used as a transfection host and for studying antigen processing and T cell recognition of class I MHC antigens.
Catalog No. 50-238-2070 Supplier ATCC Supplier No. CRL1992
T2 (174 x CEM.T2) cellls were derived from a variant of the T1 cell line (ATCC CRL-1991) produced by selection the SFR1-MI.3 monoclonal antibody (against a monomorphic determinant on HLA DR). The cell line can be used as a transfection host and for studying antigen processing and T cell recognition of class I MHC antigens.