Learn More
Sigma Aldrich T2 - thermocouple alloy, Ni95(Al+Mn+Si)5
Catalog No. 503094389 Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
50-309-4389 1 Ea.
1 options
Catalog No. 50-309-4389 Supplier Sigma Aldrich Supplier No. GF397519481EA
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
T2 - thermocouple alloy, Ni95(Al+Mn+Si)5
Specifications
Quantity 1 Ea.
Synonym NI105810