Learn More
Hach Company Radiometer Analytical T201 Temperature Sensor (glass body, cinch (RCA) plug for Radiometer PHM2xx, CDM2xx, PHM9x, IONxxx & TIMxxx meters)
Catalog No. 501997606 Shop All Hach Company Products
Encompass
The Radiometer T201 Temperature Sensor has a glass body, fixed cable, and 10 kohm resistance thermistor. The T201's cinch (RCA) connector can be used with Radiometer PHM2xx, CDM2xx, PHM9x, IONxxx and TIMxxx meters.
Catalog No. 50-199-7606 Supplier Hach Company Supplier No. E51M001
The Radiometer T201 Temperature Sensor has a glass body, fixed cable, and 10 kohm resistance thermistor. The T201's cinch (RCA) connector can be used with Radiometer PHM2xx, CDM2xx, PHM9x, IONxxx and TIMxxx meters.