Learn More
Cayman Chemical t232trIfluoroacetAT salt 10mg
SDP
Catalog No. 502911701
Encompass_Preferred
A synthetic peptide derivative of somatostatin; binds to somatostatin receptors in isolated rat synaptosomal membranes (IC50 = ~0.1 nM); inhibits the proliferation of P388 murine and HL-60 human leukemia cells at 30 and 60 UG/ml; reduces tumor volume in an HL-60 mouse xenograft model from 3-12 UG/animal per day; inhibits capsaicin-induced intradermal neutrophil accumulation and EAr edema in rats; inhibits mechano-nociceptive hyperalgesia in rats from 5-20 UG/kg, i.p.
Catalog No. 50-291-1701 Supplier Cayman Chemical Supplier No. 3604510 MG
A synthetic peptide derivative of somatostatin; binds to somatostatin receptors in isolated rat synaptosomal membranes (IC50 = ~0.1 nM); inhibits the proliferation of P388 murine and HL-60 human leukemia cells at 30 and 60 UG/ml; reduces tumor volume in an HL-60 mouse xenograft model from 3-12 UG/animal per day; inhibits capsaicin-induced intradermal neutrophil accumulation and EAr edema in rats; inhibits mechano-nociceptive hyperalgesia in rats from 5-20 UG/kg, i.p.