Learn More
Andwin Scientific T25 DIGITAL ULTRA TURRAX
SDP
Catalog No. NC2004067
Encompass_Preferred
  • High-performance dispersing instrument for homogenization, emulsification, and suspension.
  • Consistent results through electronic speed control and overload protection.
  • Wide speed range of 3000 to 25,000 rpm for versatility across different tasks.
  • Effective for a dispersing volume between 1 to 2000 ml.
  • Designed for quiet operation for uninterrupted laboratory environments.
  • Supports easily cleanable stainless steel or disposable plastic dispersing elements.
  • Digital display for precise speed control and monitoring.
  • Permissible ambient temperature range of 5 to 40 °C.
  • Protection class IP20 ensures safe operation in various conditions.
  • Lightweight at 2.5 kg for easy handling in laboratories.
Catalog No. NC2004067 Supplier Andwin Scientific Supplier No. 0003725001
  • High-performance dispersing instrument for homogenization, emulsification, and suspension.
  • Consistent results through electronic speed control and overload protection.
  • Wide speed range of 3000 to 25,000 rpm for versatility across different tasks.
  • Effective for a dispersing volume between 1 to 2000 ml.
  • Designed for quiet operation for uninterrupted laboratory environments.
  • Supports easily cleanable stainless steel or disposable plastic dispersing elements.
  • Digital display for precise speed control and monitoring.
  • Permissible ambient temperature range of 5 to 40 °C.
  • Protection class IP20 ensures safe operation in various conditions.
  • Lightweight at 2.5 kg for easy handling in laboratories.