Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC T3-ATA S-isomer 10mM 1mL | 2438721-48-1 | 1 ML
SDP

Catalog No. 5000229670
Encompass_Preferred

T3-ATA (S-isomer) is the S-isomer of T3-ATA, a thyroid-hormone analog that acts on thyroid hormone receptors. It is supplied for research use as a DMSO solution (commonly 10 mM, 1 mL) and should be stored protected from light and under inert atmosphere per manufacturer recommendations.

  • S-isomer of T3-ATA, the active form of the thyroid hormone.
  • Molecular formula: C19H16I3NO6S; molecular weight: 767.11.
  • Provided as DMSO solution; soluble in DMSO (≥ 50 mg/mL).
  • Purity indicated by COA: 99.5%.
  • Storage: 4°C protected from light; in solvent store at -80°C (6 months) or -20°C (1 month).
  • Intended for research use only.

Catalog No. 50-002-29670 Supplier Medchemexpress LLC Supplier No. HY114271A10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

T3-ATA (S-isomer) is the S-isomer of T3-ATA, a thyroid-hormone analog that acts on thyroid hormone receptors. It is supplied for research use as a DMSO solution (commonly 10 mM, 1 mL) and should be stored protected from light and under inert atmosphere per manufacturer recommendations.

  • S-isomer of T3-ATA, the active form of the thyroid hormone.
  • Molecular formula: C19H16I3NO6S; molecular weight: 767.11.
  • Provided as DMSO solution; soluble in DMSO (≥ 50 mg/mL).
  • Purity indicated by COA: 99.5%.
  • Storage: 4°C protected from light; in solvent store at -80°C (6 months) or -20°C (1 month).
  • Intended for research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.