Learn More
Qiagen Beverly Inc T3 DNA LIGASE
Catalog No. 50305903
Encompass
DNA Ligase; T3 DNA Ligase; Conc. 3,000,000 U/ml; 900,000 U; incl. 2X rapid ligation buffer
Catalog No. 50-305-903 Supplier Qiagen Beverly Inc Supplier No. L6010L
DNA Ligase; T3 DNA Ligase; Conc. 3,000,000 U/ml; 900,000 U; incl. 2X rapid ligation buffer