Learn More
Sigma Aldrich Fine Chemicals Biosciences T3 RNA Polymerase from Esc
Catalog No. 502901836
Encompass
With this enzyme homogeneously labeled single-stranded RNA can be generated. Transcripts can be nonradioactively labeled with biotin or DIG-11-UTP or radioactively labeled to high specific activity with [alpha-32P]- or [alpha-35S]-labeled nucleotides. T3 RNA polymerase is a DNA-dependent RNA polymerase that is highly specific for its promoter. Only DNA downstream from the T3 promoter will be transcribed.
Catalog No. 50-290-1836 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 11031163001
With this enzyme homogeneously labeled single-stranded RNA can be generated. Transcripts can be nonradioactively labeled with biotin or DIG-11-UTP or radioactively labeled to high specific activity with [alpha-32P]- or [alpha-35S]-labeled nucleotides. T3 RNA polymerase is a DNA-dependent RNA polymerase that is highly specific for its promoter. Only DNA downstream from the T3 promoter will be transcribed.