Learn More
Drg International Inc T3 Total Elisa
Supplier:  Drg International Inc EIA4569
Encompass_Preferred
An enzyme immunoassay for the quantitative in vitro diagnostic measurement of total T3 (triiodothyronine) in serum and plasma. Measurement of serum triiodothyronine concentration is generally regarded as a valuable tool in the diagnosis of thyroid dysfunction. The DRG T3 Total ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA), based on the principle of competitive binding. The microtiter wells are coated with a polyclonal goat-anti-mouse antibody. Standards, controls and patient serum incubate together with Assay Reagent containing monoclonal anti-T3 antibodies. 
Catalog No. 50-194-7616