Learn More
THORLABS INC T36H, IMPERIAL OPTICAL TABLE
Catalog No. NC1953756
Encompass
3' x 6' x 8.3" Imperial Optical Table 3' x 6' x 8.3" Imperial Optical Table 193 x 102 x 49 cm @ 305 kg Weight: 219.8381 Commodity: 9031.90.9160 Country of Origin: GBR RoHS: Compliant HTSU: 9031.90.9160
Catalog No. NC1953756 Supplier THORLABS INC Supplier No. T36H
3' x 6' x 8.3" Imperial Optical Table 3' x 6' x 8.3" Imperial Optical Table 193 x 102 x 49 cm @ 305 kg Weight: 219.8381 Commodity: 9031.90.9160 Country of Origin: GBR RoHS: Compliant HTSU: 9031.90.9160