Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC T4-ATA S-isomer 10mM 1mL | 2448470-97-9 | 1 ML
SDP

Catalog No. 5000228575
Encompass_Preferred

T4-ATA (S-isomer) is the S-enantiomer of the thyroid hormone analogue T4-ATA and represents the active form used in receptor studies. It is supplied for research applications and is not intended for human use.

  • S-isomer of T4-ATA, active form of the thyroid hormone.
  • CAS number 2448470-97-9.
  • Molecular formula C19H15I4NO6S; molecular weight 893.01 g/mol.
  • Purity 99.5% (as supplied).
  • Available as 10 mM solution (1 ML) in DMSO and as solid quantities (5 mg, 10 mg, 25 mg, 50 mg, 100 mg).
  • Intended for in vitro and laboratory research; not for human or clinical use.

Catalog No. 50-002-28575 Supplier Medchemexpress LLC Supplier No. HY114272A10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

T4-ATA (S-isomer) is the S-enantiomer of the thyroid hormone analogue T4-ATA and represents the active form used in receptor studies. It is supplied for research applications and is not intended for human use.

  • S-isomer of T4-ATA, active form of the thyroid hormone.
  • CAS number 2448470-97-9.
  • Molecular formula C19H15I4NO6S; molecular weight 893.01 g/mol.
  • Purity 99.5% (as supplied).
  • Available as 10 mM solution (1 ML) in DMSO and as solid quantities (5 mg, 10 mg, 25 mg, 50 mg, 100 mg).
  • Intended for in vitro and laboratory research; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.