Learn More
New England Biolabs, Inc. T4 DNA Ligase – 100,000 units – 2,000,000 units/ml
SDP
Catalog No. 50811604
Encompass
The industry standard for performance and quality. T4 DNA Ligase catalyzes the formation of a phosphodiester bond between juxtaposed 5' phosphate and 3' hydroxyl termini in duplex DNA or RNA.
  • Catalyzes the formation of a phosphodiester bond between juxtaposed 5' phosphate and 3' hydroxyl termini in duplex DNA or RNA
  • Joins blunt and cohesive end termini as well as repair single stranded nicks in duplex DNA
  • T4 DNA Ligase variants: Salt-T4, Hi-T4, Instant Sticky-end Ligase MM, Blunt/TA Ligase MM
Catalog No. 50-811-604 Supplier New England Biolabs, Inc. Supplier No. M0202M
The industry standard for performance and quality. T4 DNA Ligase catalyzes the formation of a phosphodiester bond between juxtaposed 5' phosphate and 3' hydroxyl termini in duplex DNA or RNA.
  • Catalyzes the formation of a phosphodiester bond between juxtaposed 5' phosphate and 3' hydroxyl termini in duplex DNA or RNA
  • Joins blunt and cohesive end termini as well as repair single stranded nicks in duplex DNA
  • T4 DNA Ligase variants: Salt-T4, Hi-T4, Instant Sticky-end Ligase MM, Blunt/TA Ligase MM