Learn More
INTACT GENOMICS INC T4 DNA Ligase 100,000 Units (2000 units/µl)
Catalog No. 5000082990
Encompass
Intact Genomics T4 DNA Ligase catalyzes the formation of a phosphodiester bond between juxtaposed 5’-phosphate and 3’-hydroxyl termini in duplex DNA or RNA. This enzyme joins DNA fragments with either cohesive or blunt termini and repairs single stranded nicks in duplex DNA, RNA or DNA/RNA hybrids (1).Applications• Cloning of restriction enzyme generated DNA fragments• Cloning of PCR products• Next-gen library preparation• Joining linkers and adapters to cohesive or blunt-ended DNA• Nick repair in duplex DNA, RNA or DNA/RNA hybrids• Self-circularization of linear DNA
Catalog No. 50-000-82990 Supplier INTACT GENOMICS INC Supplier No. 3216
Intact Genomics T4 DNA Ligase catalyzes the formation of a phosphodiester bond between juxtaposed 5’-phosphate and 3’-hydroxyl termini in duplex DNA or RNA. This enzyme joins DNA fragments with either cohesive or blunt termini and repairs single stranded nicks in duplex DNA, RNA or DNA/RNA hybrids (1).Applications• Cloning of restriction enzyme generated DNA fragments• Cloning of PCR products• Next-gen library preparation• Joining linkers and adapters to cohesive or blunt-ended DNA• Nick repair in duplex DNA, RNA or DNA/RNA hybrids• Self-circularization of linear DNA