Learn More
Qiagen Beverly Inc T4 DNA LIGASE (RAPID)
Catalog No. 50305905
Encompass
DNA Ligase; T4 DNA Ligase (Rapid); Conc. 600,000 U/ml; 240,000U; incl. 10X buffer and 2X rapid ligation buffer
Catalog No. 50-305-905 Supplier Qiagen Beverly Inc Supplier No. L6030HCL
DNA Ligase; T4 DNA Ligase (Rapid); Conc. 600,000 U/ml; 240,000U; incl. 10X buffer and 2X rapid ligation buffer