Learn More
Sigma Aldrich Fine Chemicals Biosciences T4 DNA Ligase recombinant
Catalog No. 502905753
Encompass
T4 DNA Ligase catalyzes the formation of phosphodiester bonds between neighboring 3-hydroxyl- and 5-phosphate ends in double-stranded DNA. Single-stranded nicks in double-stranded DNA are also closed by T4 DNA Ligase.ContentsT4 DNA Ligase supplied with 10x concentrated ligation buffer that includes ATP.
Catalog No. 50-290-5753 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 10799009001
T4 DNA Ligase catalyzes the formation of phosphodiester bonds between neighboring 3-hydroxyl- and 5-phosphate ends in double-stranded DNA. Single-stranded nicks in double-stranded DNA are also closed by T4 DNA Ligase.ContentsT4 DNA Ligase supplied with 10x concentrated ligation buffer that includes ATP.