Learn More
Sigma Aldrich Fine Chemicals Biosciences T4 DNA Ligase recombinant
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 10716359001
Encompass
T4 DNA Ligase catalyzes the formation of phosphodiester bonds between neighboring 3-hydroxyl- and 5-phosphate ends in double-stranded DNA. Single-stranded nicks in double-stranded DNA are also closed by T4 DNA Ligase.ContentsT4 DNA Ligase supplied with 10x concentrated ligation buffer that includes ATP.
Catalog No. 50-290-5744