Learn More
Apexbio Technology LLC T4 DNA Ligase (RNase-free)(Synonyms: T4 DNA Ligase, RNase-free T4 DNA Ligase, T4 Ligase, T4 DNA Joining Enzyme), 100KU.
SDP
Catalog No. 5000077822
Encompass_Preferred
T4 DNA Ligase (RNase-free) is an ATP-dependent enzyme with a molecular weight of approximately 55 3 kDa catalyzing the formation of phosphodiester bonds between adjacent 5 -phosphate and 3 -hydroxyl termini in double-stranded DNA and RNA This enzyme ligates DNA fragments possessing either cohesive or blunt ends and repairs single-strand breaks in double-stranded DNA RNA or DNA/RNA hybrids It is extensively utilized in molecular cloning linker or adaptor ligation nick-repair assays and circularization of linear DNA molecules
Catalog No. 50-000-77822 Supplier Apexbio Technology LLC Supplier No. K1100100000
T4 DNA Ligase (RNase-free) is an ATP-dependent enzyme with a molecular weight of approximately 55 3 kDa catalyzing the formation of phosphodiester bonds between adjacent 5 -phosphate and 3 -hydroxyl termini in double-stranded DNA and RNA This enzyme ligates DNA fragments possessing either cohesive or blunt ends and repairs single-strand breaks in double-stranded DNA RNA or DNA/RNA hybrids It is extensively utilized in molecular cloning linker or adaptor ligation nick-repair assays and circularization of linear DNA molecules