Learn More
New England Biolabs, Inc. T4 DNA Polymerase – 750 units
SDP
Catalog No. 50811607
Encompass
T4 DNA Polymerase catalyzes the synthesis of DNA in the 5' to 3' direction and requires the presence of template and primer. This enzyme has a 3' to 5' exonuclease activity which is much more active than that found in DNA Polymerase I (E. coli). Unlike E. coli DNA Polymerase I, T4 DNA Polymerase does not have a 5' to 3' exonuclease function.
Catalog No. 50-811-607 Supplier New England Biolabs, Inc. Supplier No. M0203L
T4 DNA Polymerase catalyzes the synthesis of DNA in the 5' to 3' direction and requires the presence of template and primer. This enzyme has a 3' to 5' exonuclease activity which is much more active than that found in DNA Polymerase I (E. coli). Unlike E. coli DNA Polymerase I, T4 DNA Polymerase does not have a 5' to 3' exonuclease function.