Learn More
INTACT GENOMICS INC T4 DNA Polymerase 500 Units
Catalog No. 5000083088
Encompass
IG™ T4 DNA Polymerase has both a DNA-dependent DNA polymerase activity and a potent 3´→5´ exonuclease activity. Applications 3´-overhang removal to form blunt ends (1, 2). 5´-overhang fill-in to form blunt ends (1, 2). Probe labeling using replacement synthesis (2). DNA library preparation for Next-generation sequencing. Ligation-independent cloning of PCR products. Second strand synthesis in site-directed mutagenesis (3).
Catalog No. 50-000-83088 Supplier INTACT GENOMICS INC Supplier No. 3222
IG™ T4 DNA Polymerase has both a DNA-dependent DNA polymerase activity and a potent 3´→5´ exonuclease activity. Applications 3´-overhang removal to form blunt ends (1, 2). 5´-overhang fill-in to form blunt ends (1, 2). Probe labeling using replacement synthesis (2). DNA library preparation for Next-generation sequencing. Ligation-independent cloning of PCR products. Second strand synthesis in site-directed mutagenesis (3).