Learn More
Medchemexpress LLC T4 DNA Polymerase T 50ug | 50ug
SDP
Catalog No. 5000194975
Encompass_Preferred
T4 DNA Polymerase a pivotal enzyme in viral DNA replication performs dual functions crucial for genome synthesis As a polymerase it synthesizes DNA and with its exonucleolytic activity it proofreads by degrading single-stranded DNA in the 3 - to 5 -direction ensuring the accuracy and fidelity of replicated viral genomic DNA T4 DNA Polymerase Protein T4 phage (His) is the recombinant T4 DNA Polymerase protein expressed by E coli with N-6 His labeled tag T4 DNA Polymerase Protein T4 phage (His) has molecular weight of 103 6 kDa
Catalog No. 50-001-94975 Supplier Medchemexpress LLC Supplier No. HYP7894050UG
T4 DNA Polymerase a pivotal enzyme in viral DNA replication performs dual functions crucial for genome synthesis As a polymerase it synthesizes DNA and with its exonucleolytic activity it proofreads by degrading single-stranded DNA in the 3 - to 5 -direction ensuring the accuracy and fidelity of replicated viral genomic DNA T4 DNA Polymerase Protein T4 phage (His) is the recombinant T4 DNA Polymerase protein expressed by E coli with N-6 His labeled tag T4 DNA Polymerase Protein T4 phage (His) has molecular weight of 103 6 kDa