Learn More
T4 GP32 Protein, Glycerol Free, G-Biosciences

Description
T4 GP32 binds cooperatively to single stranded DNA and stimulates cell-free DNA synthesis directed by the T4-encoded DNA polymerase. It improves the efficiency of reverse transcriptase during RT-PCR as well as increases the yield of PCR products.
Specifications
Specifications
| Gene ID (Entrez) | 10329603 |
| Species | E. coli |
| Product Line | Gibco |
| Protein Length | 314 amino acids including HIS tag |
| Product Type | DNA Binding Protein |
| Quantity | 1000 μg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.