Learn More
New England Biolabs, Inc. T4 Phage beta-glucosyltransferase – 500 units
SDP
Catalog No. 504383
Encompass
T4 Phage -glucosyltransferase specifically transfers the glucose moiety of uridine diphosphoglucose (UDP-Glc) to the 5-hydroxymethylcytosine (5-hmC) residues in double-stranded DNA, making beta-glucosyl-5-hydroxymethylcytosine.
Catalog No. 50-438-3 Supplier New England Biolabs, Inc. Supplier No. M0357S
T4 Phage -glucosyltransferase specifically transfers the glucose moiety of uridine diphosphoglucose (UDP-Glc) to the 5-hydroxymethylcytosine (5-hmC) residues in double-stranded DNA, making beta-glucosyl-5-hydroxymethylcytosine.