Learn More
New England Biolabs, Inc. T4 Polynucleotide Kinase – 2500 units
SDP
Catalog No. 50811601
Encompass
T4 PNK catalyzes the transfer and exchange of P from the gamma; position of ATP to the 5 -hydroxyl terminus of polynucleotides (double-and single-stranded DNA and RNA) and nucleoside 3-monophosphates. Polynucleotide Kinase also catalyzes the removal of 3-phosphoryl groups from 3-phosphoryl polynucleotides, deoxynucleoside 3-monophosphates and deoxynucleoside 3-diphosphates.
  • 5' phosphorylation of DNA/RNA for subsequent ligation
  • End labeling DNA or RNA for probes and DNA sequencing
  • Removal of 3' phosphoryl groups
Catalog No. 50-811-601 Supplier New England Biolabs, Inc. Supplier No. M0201L
T4 PNK catalyzes the transfer and exchange of P from the gamma; position of ATP to the 5 -hydroxyl terminus of polynucleotides (double-and single-stranded DNA and RNA) and nucleoside 3-monophosphates. Polynucleotide Kinase also catalyzes the removal of 3-phosphoryl groups from 3-phosphoryl polynucleotides, deoxynucleoside 3-monophosphates and deoxynucleoside 3-diphosphates.
  • 5' phosphorylation of DNA/RNA for subsequent ligation
  • End labeling DNA or RNA for probes and DNA sequencing
  • Removal of 3' phosphoryl groups