Learn More
New England Biolabs, Inc. T4 Polynucleotide Kinase (3' phosphatase minus) – 200 units
SDP
Catalog No. 50995198
Encompass
T4 Polynucleotide Kinase catalyzes the transfer and exchange of P from the gamma position of ATP to the 5 -hydroxyl terminus of double- and single-stranded DNA and RNA, as well as nucleoside 3'-monophosphates. This modified version exhibits full kinase activity with no 3' phosphatase activity.
Catalog No. 50-995-198 Supplier New England Biolabs, Inc. Supplier No. M0236S
T4 Polynucleotide Kinase catalyzes the transfer and exchange of P from the gamma position of ATP to the 5 -hydroxyl terminus of double- and single-stranded DNA and RNA, as well as nucleoside 3'-monophosphates. This modified version exhibits full kinase activity with no 3' phosphatase activity.