Learn More
LGC GENOMICS LLC (Lucigen) T4 Polynucleotide Kinase, Cloned, 3,000 U @ 10 U/LAn enzyme for the phosphorylation 5' hydroxy ends of ssDNA and dsDNA, RNA, and nucleoside 3 monophosphates.
Catalog No. NC2162072
Encompass
Phosphorylate 5' hydroxy ends of ssDNA and dsDNA, RNA, and nucleoside 3 monophosphatesHighly active enzyme that readily phosphorylates 5' hydroxyl ends of nucleic acidsFlexible: Use newly phosphorylated nucleic acids in a variety of applications such as cloning, next-gen sequencing library prep, and preparation of labelled nucleic acidsEach T4 Polynucleotide Kinase includes the enzyme and 10X Reaction Buffer without ATP. ATP is available separately.
Catalog No. NC2162072 Supplier LGC GENOMICS LLC Supplier No. P0503K
Phosphorylate 5' hydroxy ends of ssDNA and dsDNA, RNA, and nucleoside 3 monophosphatesHighly active enzyme that readily phosphorylates 5' hydroxyl ends of nucleic acidsFlexible: Use newly phosphorylated nucleic acids in a variety of applications such as cloning, next-gen sequencing library prep, and preparation of labelled nucleic acidsEach T4 Polynucleotide Kinase includes the enzyme and 10X Reaction Buffer without ATP. ATP is available separately.