Learn More
New England Biolabs, Inc. T4 RNA Ligase 1 (ssRNA Ligase) – 5000 units
SDP
Catalog No. 50811609
Encompass
T4 RNA Ligase 1 catalyzes the ligation of a 5' phosphoryl-terminated nucleic acid donor to a 3' hydroxyl-terminated nucleic acid acceptor through the formation of a 3' - 5' phosphodiester bond with hydrolysis of ATP to AMP and PPi. Substrates include single-stranded RNA and DNA as well as dinucleoside pyrophosphates.
Catalog No. 50-811-609 Supplier New England Biolabs, Inc. Supplier No. M0204L
T4 RNA Ligase 1 catalyzes the ligation of a 5' phosphoryl-terminated nucleic acid donor to a 3' hydroxyl-terminated nucleic acid acceptor through the formation of a 3' - 5' phosphodiester bond with hydrolysis of ATP to AMP and PPi. Substrates include single-stranded RNA and DNA as well as dinucleoside pyrophosphates.