Learn More
New England Biolabs, Inc. T4 RNA Ligase 2 – 150 units
SDP
Catalog No. 50995201
Encompass
T4 RNA Ligase 2, also known as T4 Rnl2 (gp24.1), has both intermolecular and intramolecular RNA strand joining activity). Unlike T4 RNA Ligase 1 , T4 RNA Ligase 2 is much more active joining nicks on double stranded RNA than on joining the ends of single stranded RNA. The enzyme requires an adjacent 5 phosphate and 3' OH for ligation. The enzyme can also ligate the 3' OH of RNA to the 5 phosphate of DNA in a double stranded structure.
Catalog No. 50-995-201 Supplier New England Biolabs, Inc. Supplier No. M0239S
T4 RNA Ligase 2, also known as T4 Rnl2 (gp24.1), has both intermolecular and intramolecular RNA strand joining activity). Unlike T4 RNA Ligase 1 , T4 RNA Ligase 2 is much more active joining nicks on double stranded RNA than on joining the ends of single stranded RNA. The enzyme requires an adjacent 5 phosphate and 3' OH for ligation. The enzyme can also ligate the 3' OH of RNA to the 5 phosphate of DNA in a double stranded structure.