Learn More
AMID BIOSCIENCES Amid Biosciences, T4 RNA Ligase 2, Recombinant, RNA Ligation, 1000 Units
SDP
Catalog No. NC4043436
Encompass_Preferred
T4 RNA Ligase 2 is supplied in a 1000 Units format for RNA ligation workflows that require enzymatic joining of RNA substrates or adapter containing molecules. Applications include Intermolecular and intramolecular RNA strand joining, nick ligation on double stranded RNA, RNA-DNA hybrid ligation, ligation of 3 OH RNA to 5 phosphate DNA in duplex structures. This product includes 1mL of 10X Reaction Buffer, 100 mM Tris-HCl, pH 7.5, 20 mM MgCl2, 20 mM DTT, 5 mM ATP. Requires ATP unless the substrate is preadenylated. Technical specifications are included below.- Quantity - 1000 Units- Organism - Bacteriophage T4- Activity - 10,000 U/mL (greater than 40,000 U/mg)- Buffer - 10 mM Tris-HCl, pH 7.5, 100 mM NaCl, 0.1 mM DTT, 0.1 mM EDTA, 50% Glycerol- Storage - -20 CThis product is for research use only.
Catalog No. NC4043436 Supplier AMID BIOSCIENCES Supplier No. RNL23012
T4 RNA Ligase 2 is supplied in a 1000 Units format for RNA ligation workflows that require enzymatic joining of RNA substrates or adapter containing molecules. Applications include Intermolecular and intramolecular RNA strand joining, nick ligation on double stranded RNA, RNA-DNA hybrid ligation, ligation of 3 OH RNA to 5 phosphate DNA in duplex structures. This product includes 1mL of 10X Reaction Buffer, 100 mM Tris-HCl, pH 7.5, 20 mM MgCl2, 20 mM DTT, 5 mM ATP. Requires ATP unless the substrate is preadenylated. Technical specifications are included below.- Quantity - 1000 Units- Organism - Bacteriophage T4- Activity - 10,000 U/mL (greater than 40,000 U/mg)- Buffer - 10 mM Tris-HCl, pH 7.5, 100 mM NaCl, 0.1 mM DTT, 0.1 mM EDTA, 50% Glycerol- Storage - -20 CThis product is for research use only.