Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedical™ T4 (Total) ChLIA Kit

Catalog No. MP07M275B Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 x 96 Wells
2 x 96 Wells
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP07M275B 2 x 96 Wells
MP07M275A 1 x 96 Wells
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP07M275B Supplier MP Biomedical™ Supplier No. 07M275B
Add to Cart
Add to Cart

Quantitative Chemiluminescence Immunoassay

The T4 (Total) Chemiluminescence Immunoassay kit is intended for the measurement of Total Thyroxine (T4) in human serum or plasma.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Kit and Assay
Sample Type Plasma, Serum
Target Species Human
Quantity 2 x 96 Wells
Sample Volume 25 μL
Storage Requirements 2°C to 8°C
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.