Learn More
INTACT GENOMICS INC T4 UvsX DNA Recombinase 1000µg
Catalog No. 5000083021
Encompass
T4 UvsX DNA Recombinase is useful for Recombinase Polymerase Amplification (RPA) and other applications.Homologous recombination is important for the error-free repair of DNA double-strand breaks and for replication fork restart. Recombinases of the RecA/RAD51 family perform the central catalytic role in this process. UvsX recombinase is the RecA/Rad51 ortholog of bacteriophage T4. T4 UvsX DNA Recombinase and other recombinases form presynaptic filaments on ssDNA that are activated to search for homology in dsDNA and to perform DNA strand exchange (1-3).Product Components 1) UvsX Recombinase 2) 10X UvsX Recombinase Reaction Buffer1x UvsX Recombinase Reaction Buffer Composition:20 mM Tris-acetate pH 7.8100 mM Potassium acetate10 mM Magnesium acetate1 mM DTTStorage Temperature: –20ºC
Catalog No. 50-000-83021 Supplier INTACT GENOMICS INC Supplier No. 3567
T4 UvsX DNA Recombinase is useful for Recombinase Polymerase Amplification (RPA) and other applications.Homologous recombination is important for the error-free repair of DNA double-strand breaks and for replication fork restart. Recombinases of the RecA/RAD51 family perform the central catalytic role in this process. UvsX recombinase is the RecA/Rad51 ortholog of bacteriophage T4. T4 UvsX DNA Recombinase and other recombinases form presynaptic filaments on ssDNA that are activated to search for homology in dsDNA and to perform DNA strand exchange (1-3).Product Components 1) UvsX Recombinase 2) 10X UvsX Recombinase Reaction Buffer1x UvsX Recombinase Reaction Buffer Composition:20 mM Tris-acetate pH 7.8100 mM Potassium acetate10 mM Magnesium acetate1 mM DTTStorage Temperature: –20ºC