Learn More
Electronic Imaging Materials Inc 4.33 Inches x 74 Meter T44 Premium Wax/Resin Ribbon - EIMINC
SDP
Catalog No. NC1430605
Encompass_Preferred
T44—This 4.33 inches x 74 meter premium all-purpose wax/resin formula is designed to print on a wide variety of materials,offering excellent barcode scanning with sharp, reliable images printed at a wide variety of print speeds. Great chemical resistance when used with 651 XyResistslide labels.*Note: make sure to find out if ribbon is compatible with your printer & label material before purchasing. Works in Zebra GX430 series printers, as well as TSC TC 200/210/300/310 printers.
Catalog No. NC1430605 Supplier Electronic Imaging Materials Inc Supplier No. T44433074PCQTY 611
T44—This 4.33 inches x 74 meter premium all-purpose wax/resin formula is designed to print on a wide variety of materials,offering excellent barcode scanning with sharp, reliable images printed at a wide variety of print speeds. Great chemical resistance when used with 651 XyResistslide labels.*Note: make sure to find out if ribbon is compatible with your printer & label material before purchasing. Works in Zebra GX430 series printers, as well as TSC TC 200/210/300/310 printers.