Learn More
ATCC T47D-KBluc; Breast Ductal Carcinoma; Human (Homo sapiens)
Catalog No. 502384204
Encompass
T47D-KBluc is an epithelial cell line that was isolated in 2000 from a pleural effusion of a 54-year-old, female patient with ductal carcinoma of the mammary gland. This cell line was deposited by VS Wilson and can be used in assay development and cancer research.
Catalog No. 50-238-4204 Supplier ATCC Supplier No. CRL2865
T47D-KBluc is an epithelial cell line that was isolated in 2000 from a pleural effusion of a 54-year-old, female patient with ductal carcinoma of the mammary gland. This cell line was deposited by VS Wilson and can be used in assay development and cancer research.