Learn More
New England Biolabs, Inc. T5 Exonuclease – 1000 units
SDP
Catalog No. NC1872798
Encompass
T5 Exonuclease degrades DNA in the 5 to 3 direction. T5 Exonuclease is able to initiate nucleotide removal from the 5 termini or at gaps and nicks of linear or circular dsDNA. However, the enzyme does not degrade supercoiled dsDNA. T5 Exonuclease also has ssDNA endonuclease activity.
Catalog No. NC1872798 Supplier New England Biolabs, Inc. Supplier No. M0663S
T5 Exonuclease degrades DNA in the 5 to 3 direction. T5 Exonuclease is able to initiate nucleotide removal from the 5 termini or at gaps and nicks of linear or circular dsDNA. However, the enzyme does not degrade supercoiled dsDNA. T5 Exonuclease also has ssDNA endonuclease activity.