Learn More
Alkali Scientific T5000-96-2D TC 96 Turbo 2D Gra
SDP
Catalog No. 503029617
Encompass_Preferred
The Alkali TC-96 Turbo 2D Gradient Thermal Cycler is designed for fast cycling and quick protocol optimization Exceptionally easy to program the unit features tight temperature control and excellent block uniformity for consistent reliable results Many thermal cyclers offer users the option to select a temperature gradient across the block to test a range of annealing temperatures simultaneously The TC-96 Turbo offers a 2D gradient across both the columns and rows allowing for optimization of annealing and denaturation temperatures in the same run The ability to optimize both of these steps provides for an increase in yield and specificity of complex or GC rich templates Temperature ramp rates as fast as 9C per second allow reactions to be performed quickly up to 20% faster than other popular thermal cyclers Temperature change is precisely controlled by a proprietary algorithm that simulates sample temperature and allows samples to arrive at the programmed temperature without
Catalog No. 50-302-9617 Supplier Alkali Scientific Supplier No. T5000962D
Add to Cart
Add to Cart
Request a Quote
The Alkali TC-96 Turbo 2D Gradient Thermal Cycler is designed for fast cycling and quick protocol optimization Exceptionally easy to program the unit features tight temperature control and excellent block uniformity for consistent reliable results Many thermal cyclers offer users the option to select a temperature gradient across the block to test a range of annealing temperatures simultaneously The TC-96 Turbo offers a 2D gradient across both the columns and rows allowing for optimization of annealing and denaturation temperatures in the same run The ability to optimize both of these steps provides for an increase in yield and specificity of complex or GC rich templates Temperature ramp rates as fast as 9C per second allow reactions to be performed quickly up to 20% faster than other popular thermal cyclers Temperature change is precisely controlled by a proprietary algorithm that simulates sample temperature and allows samples to arrive at the programmed temperature without