Learn More
Alkali Scientific T5000-96 TC 9639 Thermal Cycle
SDP
Catalog No. 503029595
Encompass_Preferred
Alkalis T5000-96 TC9639 Thermal Cycler provides consistent reliable results for laboratories with medium to high throughput From its large touch-screen user interface to its unique multi-format block this cycler is loaded with features The program wizard and 10 preinstalled programs make it easier than ever to get up and running A large full color touch screen serves as the operations center where all functions are accessed From the Home screen simply touch an icon to view the last run program enter the program library or use the program wizard to quickly enter a protocol In addition icons for favorite programs can be added for faster access The program wizard guides users through quickly setting up a protocol in a single screen Once entered programs are shown graphically and can be edited by simply touching the step that requires modification Time and temperature increments can be built into programs as well as slower ramp rates Storage capabilities are unlimited programs can
Catalog No. 50-302-9595 Supplier Alkali Scientific Supplier No. T500096
Add to Cart
Add to Cart
Request a Quote
Alkalis T5000-96 TC9639 Thermal Cycler provides consistent reliable results for laboratories with medium to high throughput From its large touch-screen user interface to its unique multi-format block this cycler is loaded with features The program wizard and 10 preinstalled programs make it easier than ever to get up and running A large full color touch screen serves as the operations center where all functions are accessed From the Home screen simply touch an icon to view the last run program enter the program library or use the program wizard to quickly enter a protocol In addition icons for favorite programs can be added for faster access The program wizard guides users through quickly setting up a protocol in a single screen Once entered programs are shown graphically and can be edited by simply touching the step that requires modification Time and temperature increments can be built into programs as well as slower ramp rates Storage capabilities are unlimited programs can