Learn More
Alkali Scientific T5005-3205 TC-32 Mini Thermal
SDP
Catalog No. 503029563
Encompass_Preferred
Mini sized 9 x 7 inch footprint Capacity for 32 x 0 2ml or 4 PCR Strips Intuitive touch screen control Program Wizard with familiar icon driven programming Optimize results with Alkali ScientificPCR reagents Alkalis TC-32 Mini Thermal Cycler retains the advanced programming features easy to use software and fast reliable cycling of the larger TC-9639 Thermal Cycler but with a smaller block and lower price point Designed for lower throughput needs the TC-32 Mini Thermal Cycler has a capacity for 320 2ml tubes The wells are arranged in a 48 format allowing the block to accept strips of 8 as well as individual tubes The block heats and cools very quickly at rates as fast as 5C/second A proprietary algorithm controls block temperature bringing samples quickly to the target temperature without any over- or under-shoot Excellent block uniformity ensuresconsistency sample to sample Operation of the thermal cycler is controlled from the large full color touch screen Icon driven menus
Catalog No. 50-302-9563 Supplier Alkali Scientific Supplier No. RS50053205
Mini sized 9 x 7 inch footprint Capacity for 32 x 0 2ml or 4 PCR Strips Intuitive touch screen control Program Wizard with familiar icon driven programming Optimize results with Alkali ScientificPCR reagents Alkalis TC-32 Mini Thermal Cycler retains the advanced programming features easy to use software and fast reliable cycling of the larger TC-9639 Thermal Cycler but with a smaller block and lower price point Designed for lower throughput needs the TC-32 Mini Thermal Cycler has a capacity for 320 2ml tubes The wells are arranged in a 48 format allowing the block to accept strips of 8 as well as individual tubes The block heats and cools very quickly at rates as fast as 5C/second A proprietary algorithm controls block temperature bringing samples quickly to the target temperature without any over- or under-shoot Excellent block uniformity ensuresconsistency sample to sample Operation of the thermal cycler is controlled from the large full color touch screen Icon driven menus