Learn More
TARGETMOL CHEMICALS INC T521 5MG
SDP
Catalog No. 502580480
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. T521 is a selective inhibitor of the PBD of Plk1 and shows no inhibitory effect on Plk2 and Plk3. purity: 99%
Catalog No. 50-258-0480 Supplier TARGETMOL CHEMICALS INC Supplier No. T289045MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. T521 is a selective inhibitor of the PBD of Plk1 and shows no inhibitory effect on Plk2 and Plk3. purity: 99%