Learn More
Selleck Chemical LLC T56-LIMKi
SDP
Catalog No. 5000002952
Encompass_Preferred
T56-LIMKi is a selective inhibitor of LIMK2
Catalog No. 50-000-02952 Supplier Selleck Chemical LLC Supplier No. S64985MG
T56-LIMKi is a selective inhibitor of LIMK2