Learn More
Medchemexpress LLC HY-19352 5mg Medchemexpress, T56-LIMKi CAS:924473-59-6 Purity:>98%
SDP
Catalog No. 502030850
Encompass_Preferred
Medchemexpress, HY-19352 5mg T56-LIMKi CAS:924473-59-6 T56-LIMKi is a selective inhibitor of LIMK2; inhibits the growth of Panc-1 cells with an IC50 of 35.2 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0850 Supplier Medchemexpress LLC Supplier No. HY193525MG
Medchemexpress, HY-19352 5mg T56-LIMKi CAS:924473-59-6 T56-LIMKi is a selective inhibitor of LIMK2; inhibits the growth of Panc-1 cells with an IC50 of 35.2 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.