Learn More
Aobious Inc 4-(3-Bromophenyl)sulfonyl-N-(1-thiophen-2-ylethyl)piperazine-1-carboxamide CAS No: 1090528-71-4. Molecular Weight 458.39. Purity 98% by HPLC
Catalog No. NC1407886
Encompass
Novel inhibitor of MyD88-dependent signaling pathways, disrupting MyD88 homodimeric formation, which is critical for its signaling function. Storage Condition 0°C (short term), -20°C (long term), desiccated
Catalog No. NC1407886 Supplier Aobious Inc Supplier No. AOB58751MG
Novel inhibitor of MyD88-dependent signaling pathways, disrupting MyD88 homodimeric formation, which is critical for its signaling function. Storage Condition 0°C (short term), -20°C (long term), desiccated