Learn More
Thermo Scientific™ pT7CFE1-NHis-GST-CHA Vector for Mammalian Cell-Free Protein Expression
Description
Thermo Scientific™ T7 Cell-Free Expression Vectors (pT7CFE1) are cloning plasmids optimized to use with the Thermo Scientific 1-Step Human In Vitro Protein Expression System for in vitro translation (IVT) of tagged fusion proteins.
Highlights:
Optimized performance – designed to provide the highest yield in the human in vitro translation system
Many options – multiple tag and tag-location options available
Modular MCS – multiple cloning site is maintained across vector family to facilitate subcloning
Cleavable tags – HRV 3C cleavage site available on select vectors
pT7CFE1 Plasmid Features:
- EMCV IRES at the 5-prime UTR promotes high-level translation of mRNAs
- MCS accommodates gene insertion via ten different restriction sites: Msc1, Nde1, BamH1, EcoR1, EcoRV, Pac1, Pst1, Sac1, Sal1, Not1 and Xho1
- Poly A sequence in the 3-prime region promotes mRNA stabilization and protection from nucleases
- T7 terminator ensures synthesis of accurate size mRNA transcripts
- Plasmid linearization may be accomplished with restriction sites between Poly A sequence and the T7 terminator region
Specifications
Specifications
| Description | pT7CFE1-NHis-GST-CHA |
| Type | pT7CFE1-NHis-GST-CHA Vector |
| Cloning Method | Restriction Enzyme/MCS |
| Protein Tag | GST Tag, His Tag, HA Tag |
| Quantity | 10 μg |
| Content And Storage | Store between -70°C and -20°C. |
| Antibiotic Resistance Bacterial | Ampicillin (AmpR) |
| Promoter | T7 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.