Learn More
ARBOR BIOSCIENCES T7 deGFP Control Plasmid 25ul
Catalog No. NC3673210
Encompass
Standalone version of the T7 deGFP Control Plasmid that is provided with the myTXTL Pro Kit. For optimizing reaction setup. 24 nM concentration.
Catalog No. NC3673210 Supplier ARBOR BIOSCIENCES Supplier No. 503002
Standalone version of the T7 deGFP Control Plasmid that is provided with the myTXTL Pro Kit. For optimizing reaction setup. 24 nM concentration.