Learn More
Qiagen Beverly Inc T7 DNA LIGASE
Catalog No. 50305909
Encompass
DNA Ligase; T7 DNA Ligase; Conc. 3,000,000 U/ul; 900,000 U; incl. 2x Rapid Ligation buffer
Catalog No. 50-305-909 Supplier Qiagen Beverly Inc Supplier No. L6020L
DNA Ligase; T7 DNA Ligase; Conc. 3,000,000 U/ul; 900,000 U; incl. 2x Rapid Ligation buffer