Learn More
New England Biolabs, Inc. T7 DNA Polymerase (unmodified) – 1500 units
SDP
Catalog No. 50995208
Encompass
T7 DNA Polymerase catalyzes the replication of T7 phage DNA during infection. The protein dimer has two catalytic activities: DNA polymerase activity and strong 3 5 exonuclease. The high fidelity and rapid extension rate of the enzyme make it particularly useful in copying long stretches of DNA template.
Catalog No. 50-995-208 Supplier New England Biolabs, Inc. Supplier No. M0274L
T7 DNA Polymerase catalyzes the replication of T7 phage DNA during infection. The protein dimer has two catalytic activities: DNA polymerase activity and strong 3 5 exonuclease. The high fidelity and rapid extension rate of the enzyme make it particularly useful in copying long stretches of DNA template.