Learn More
New England Biolabs, Inc. T7 Endonuclease I – 1250 units
SDP
Catalog No. 50995224
Encompass
T7 Endonuclease I recognizes and cleaves non-perfectly matched DNA, cruciform DNA structures, Holliday structures or junctions, heteroduplex DNA and more slowly, nicked double-stranded DNA. The cleavage site is at the first, second or third phosphodiester bond that is 5' to the mismatch. The protein is the product of T7 gene 3.
Catalog No. 50-995-224 Supplier New England Biolabs, Inc. Supplier No. M0302L
T7 Endonuclease I recognizes and cleaves non-perfectly matched DNA, cruciform DNA structures, Holliday structures or junctions, heteroduplex DNA and more slowly, nicked double-stranded DNA. The cleavage site is at the first, second or third phosphodiester bond that is 5' to the mismatch. The protein is the product of T7 gene 3.