Learn More
New England Biolabs, Inc. T7 Exonuclease – 5000 units
SDP
Catalog No. 50811688
Encompass
T7 Exonuclease acts in the 5' to 3' direction, catalyzing the removal of 5' mononucleotides from linear or nicked duplex DNA.
Catalog No. 50-811-688 Supplier New England Biolabs, Inc. Supplier No. M0263L
T7 Exonuclease acts in the 5' to 3' direction, catalyzing the removal of 5' mononucleotides from linear or nicked duplex DNA.