Learn More
New England Biolabs, Inc. T7 Express lysY Competent E. coli (High Efficiency) – 6 x 0.2 ml/tube
SDP
Catalog No. 50995057
Encompass
High efficiency chemically competent E. coli cells suitable for T7 expression with enhanced reduction of basal expression.
Catalog No. 50-995-057 Supplier New England Biolabs, Inc. Supplier No. C3010I
High efficiency chemically competent E. coli cells suitable for T7 expression with enhanced reduction of basal expression.