Learn More
New England Biolabs, Inc. HiScribe™ T7 High Yield RNA Synthesis Kit – 50 reactions
SDP
Catalog No. 504299
Encompass
The HiScribe T7 High Yield RNA Synthesis Kit is an extremely flexible system for in vitro transcription of RNA using T7 RNA Polymerase. The kit allows for synthesis of many kinds of RNA including internally labeled and co-transcriptionally capped transcripts. RNA synthesized from the kit is suitable for many applications including RNA structure and function studies, ribozyme biochemistry, probes for RNase protection assays and hybridization based blots, anti-sense RNA and RNAi experiments, microarray analysis, microinjection, and in vitro translation and RNA vaccines. The kit contains sufficient reagents for 50 reactions of 20 µl each. Each standard reaction yields up to 180 µg of RNA from 1 µg control template. Each kit can yield up to 9 mg RNA. For 32P labeling, the kit contains enough reagents for 100 reactions of 20 µl each.
  • Linearized control template included for verification of RNA synthesis
  • Includes separate tube of DTT
Catalog No. 50-429-9 Supplier New England Biolabs, Inc. Supplier No. E2040S
The HiScribe T7 High Yield RNA Synthesis Kit is an extremely flexible system for in vitro transcription of RNA using T7 RNA Polymerase. The kit allows for synthesis of many kinds of RNA including internally labeled and co-transcriptionally capped transcripts. RNA synthesized from the kit is suitable for many applications including RNA structure and function studies, ribozyme biochemistry, probes for RNase protection assays and hybridization based blots, anti-sense RNA and RNAi experiments, microarray analysis, microinjection, and in vitro translation and RNA vaccines. The kit contains sufficient reagents for 50 reactions of 20 µl each. Each standard reaction yields up to 180 µg of RNA from 1 µg control template. Each kit can yield up to 9 mg RNA. For 32P labeling, the kit contains enough reagents for 100 reactions of 20 µl each.
  • Linearized control template included for verification of RNA synthesis
  • Includes separate tube of DTT